Skip to Content
Merck
All Photos(1)

Key Documents

391735

Sigma-Aldrich

1-Hydroxy-6-(trifluoromethyl)benzotriazole

98%

Synonym(s):

6-(Trifluoromethyl)-HOBT

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H4F3N3O
CAS Number:
Molecular Weight:
203.12
Beilstein:
614848
MDL number:
UNSPSC Code:
12352200

Assay

98%

mp

143-145 °C (lit.)

SMILES string

On1nnc2ccc(cc12)C(F)(F)F

Looking for similar products? Visit Product Comparison Guide

Application

Applications include the preparation of Ogura′s BTBO[1] and BTBC[2] coupling agents.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis, 557-557 (1987)
The Journal of Organic Chemistry, 50, 273-273 (1985)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service