Skip to Content
Merck
All Photos(1)

Documents

390895

Sigma-Aldrich

Ethyl 3,5-dichloro-4-hydroxybenzoate hydrate

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Cl2C6H2(OH)CO2C2H5·xH2O
CAS Number:
Molecular Weight:
235.06 (anhydrous basis)
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

98%

mp

110-115 °C (lit.)

SMILES string

O.CCOC(=O)c1cc(Cl)c(O)c(Cl)c1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service