Skip to Content
Merck
All Photos(1)

Documents

390674

Sigma-Aldrich

5-(Trifluoromethoxy)isatin

98%

Synonym(s):

5-(Trifluoromethoxy)indoline-2,3-dione

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H4F3NO3
CAS Number:
Molecular Weight:
231.13
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

98%

mp

170-172 °C (lit.)

SMILES string

FC(F)(F)Oc1ccc2NC(=O)C(=O)c2c1

InChI

1S/C9H4F3NO3/c10-9(11,12)16-4-1-2-6-5(3-4)7(14)8(15)13-6/h1-3H,(H,13,14,15)

InChI key

XHAJMVPMNOBILF-UHFFFAOYSA-N

Application

Reactant for preparation of (fluorinated-indolinonylidene)-hydrazine-carbothioamides by condensation with thiosemicarbazides
Reactant for preparation of oxindole derivatives as TAk1 kinase inhibitors
Reactant for preparation of isatin thiosemicarbazones via condensation with thiosemicarbazones as herpes simplex virus (HSV) inhibitors
Reactant for synthesis of ortho disubstituted biphenyls
Reactant for preparation of indolinon-spirothiazolidinones as selective inhibitors of Mycobacterium tuberculosis protein tyrosine phosphatase B
Reactant for synthesis of 3-substituted 2-indolinone RET kinase inhibitors
Reactant for preparation of indolone acetamides as antiseizure agents

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service