387991
Poly(vinyl chloride-co-vinyl acetate-co-maleic acid)
average Mn ~22,000
About This Item
Recommended Products
mol wt
average Mn ~22,000
inherent viscosity
0.50(lit.)
acid number
10 mg KOH/g
transition temp
Tg 74 °C
density
1.35 g/mL at 25 °C (lit.)
SMILES string
ClC=C.CC(=O)OC=C.OC(=O)\C=C\C(O)=O
Looking for similar products? Visit Product Comparison Guide
Warning
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service