Skip to Content
Merck
All Photos(1)

Documents

38784

Sigma-Aldrich

4-[(2,4-Dimethoxyphenyl)(Fmoc-amino)methyl]phenoxyacetic acid

≥99.0% (HPLC)

Synonym(s):

Rink amide linker

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C32H29NO7
CAS Number:
Molecular Weight:
539.58
Beilstein:
5841553
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.22

Assay

≥99.0% (HPLC)

form

solid

reaction suitability

reagent type: cross-linking reagent

mp

~180 °C (dec.)

functional group

Fmoc
amine
carboxylic acid

storage temp.

2-8°C

SMILES string

O=C(O)COC1=CC=C(C=C1)C(NC(OCC2C(C=CC=C3)=C3C4=C2C=CC=C4)=O)C5=CC=C(OC)C=C5OC

InChI

1S/C32H29NO7/c1-37-22-15-16-27(29(17-22)38-2)31(20-11-13-21(14-12-20)39-19-30(34)35)33-32(36)40-18-28-25-9-5-3-7-23(25)24-8-4-6-10-26(24)28/h3-17,28,31H,18-19H2,1-2H3,(H,33,36)(H,34,35)

InChI key

UPMGJEMWPQOACJ-UHFFFAOYSA-N

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hiroshi Hinou et al.
Bioorganic & medicinal chemistry, 27(13), 2822-2831 (2019-05-14)
Structural and functional effects of core M1 type glycan modification catalyzed by protein O-linked mannose β1,2-N-acetylglucosaminyltransferase 1 (POMGnT1) were investigated using a core M1 glycoform focused library of an α-dystroglycan fragment, 372TRGAIIQTPTLGPIQPTRV390. Evanescent-field fluorescence-assisted microarray system illuminated the specific binding
Thomas H Craven et al.
Scientific reports, 8(1), 13490-13490 (2018-09-12)
Serine proteases are released by neutrophils to act primarily as antimicrobial proteins but excessive and unbalanced serine protease activity results in serious host tissue damage. Here the synthesis of a novel chemical sensor based on a multi-branched fluorescence quencher is
Alessandra Soriente et al.
Journal of materials science. Materials in medicine, 29(5), 62-62 (2018-05-08)
The present work is focused on the design of a bioactive chitosan-based scaffold functionalized with organic and inorganic signals to provide the biochemical cues for promoting stem cell osteogenic commitment. The first approach is based on the use of a
Jamin B Hein et al.
Bio-protocol, 10(13), e3669-e3669 (2021-03-05)
Every living cell relies on signal transduction pathways comprised of protein-protein interactions (PPIs). In many cases, these PPIs are between a folded protein domain and a short linear motif (SLiM) within an unstructured region of a protein. As a result
Lubinda Mbundi et al.
Nanomaterials (Basel, Switzerland), 8(6) (2018-06-06)
The control of osteoblast/osteoclast cross-talk is crucial in the bone remodelling process and provides a target mechanism in the development of drugs for bone metabolic diseases. Osteoprotegerin is a key molecule in this biosignalling pathway as it inhibits osteoclastogenesis and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service