Skip to Content
Merck
All Photos(1)

Key Documents

376655

Sigma-Aldrich

Diisooctyl phthalate

ester content ≥99 % (mixture of C8 isomers), technical grade

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H4-1,2-(CO2C8H17)2
CAS Number:
Molecular Weight:
390.56
EC Number:
UNSPSC Code:
12162002
PubChem Substance ID:

grade

technical grade

vapor pressure

1 mmHg ( 200 °C)

composition

ester content, ≥99% (mixture of C8 isomers)

impurities

≤2% dioctyl phthalate

refractive index

n20/D 1.486 (lit.)

density

0.983 g/mL at 25 °C (lit.)

SMILES string

CCCCCCCCOC(=O)c1ccccc1C(=O)OCCCCCCCC

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Repr. 1B

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 1


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH SVHC Candidate List

CAS No.

EU REACH Annex XVII (Restriction List)

CAS No.

EU REACH Annex XIV (Authorisation List)

CAS No.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

L Cherta et al.
Food chemistry, 188, 301-308 (2015-06-05)
The potential of an advanced analytical strategy based on the use of gas chromatography (GC) coupled to high resolution mass spectrometry (HRMS) with two different analyzers and ionization sources has been investigated and applied to the non-target analysis of food

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service