Skip to Content
Merck
All Photos(1)

Key Documents

371599

Sigma-Aldrich

1-(4-Hydroxyphenyl)-1H-tetrazole-5-thiol

97%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H6N4OS
CAS Number:
Molecular Weight:
194.21
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

97%

mp

170 °C (dec.) (lit.)

solubility

methanol: soluble 25 mg/mL, clear (colorless to pink to purple)

SMILES string

Oc1ccc(cc1)-n2nnnc2S

InChI

1S/C7H6N4OS/c12-6-3-1-5(2-4-6)11-7(13)8-9-10-11/h1-4,12H,(H,8,10,13)

InChI key

MOXZSKYLLSPATM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

1-(4-Hydroxyphenyl)-1H-tetrazole-5-thiol is a heterocyclic thionate. Iodimetric determination of 1-(4-hydroxyphenyl)-1H-tetrazole-5-thiol in alkaline medium, via volumetric titration with potentiometric end-point detection is reported. Coordination chemistry of 1-(4-hydroxyphenyl)-1H-tetrazole-5-thiol as ligand, having deprotonated heterocyclic thioamide group, has been reported.

Application

1-(4-Hydroxyphenyl)-1H-tetrazole-5-thiol may be used in the synthesis of organotin(IV) complexes.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Syntheses and Structural Characterization of Triorganotin Derivatives of 3-Mercapto-4-methyl-4H-1, 2, 4-triazol:
Ma C, et al.
Journal of Inorganic and Organometallic Polymers and Materials, 16(2), 139-145 (2006)
Hydrogen bonding and p-p stacking interaction in the coordination of sulfur and nitrogen donor heterocycle to organotin (IV): syntheses and crystal structures of di-and triorganotin (IV) derivatives with 1-(4-hydroxyphenyl)-1< i> H</i>-tetrazole-5-thiol.
Ma C and Sun J.
Polyhedron, 23(9), 1547-1555 (2004)
Iodimetric titration of sulfur compounds in alkaline medium.
Ciesielski W and Zakrzewski R.
Chem. Anal. (Warsaw), 51(5), 653-653 (2006)
Potentiometric titration of triazolethiols and tetrazolethiols with iodine in alkaline medium.
Ciesielski W and Krenc A.
Collection of Czechoslovak Chemical Communications, 67(8), 1193-1199 (2002)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service