Skip to Content
Merck
All Photos(1)

Key Documents

365459

Sigma-Aldrich

3-(Dimethylamino)-2-methyl-2-propenal

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2NCH=C(CH3)CHO
CAS Number:
Molecular Weight:
113.16
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

97%

form

solid

bp

110-115 °C/1 mmHg (lit.)

mp

34-38 °C (lit.)

density

0.971 g/mL at 25 °C (lit.)

storage temp.

2-8°C

SMILES string

CN(C)\C=C(/C)C=O

InChI

1S/C6H11NO/c1-6(5-8)4-7(2)3/h4-5H,1-3H3/b6-4+

InChI key

JGAMOQYFDMQPRJ-GQCTYLIASA-N

General description

Photophysics of 3-dimethylamino-2-methyl-propenal (DMAMP) on excitation to the S2 (ΠΠ*) electronic state using the resonance Raman spectroscopy is reported.

Application

3-(Dimethylamino)-2-methyl-2-propenal is suitable for use in the synthesis of methylmalondialdehyde.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Validation of the Analytical Procedure for the Determination of Malondialdehyde and Three Other Aldehydes in Vegetable Oil Using Liquid Chromatography Coupled to Tandem Mass Spectrometry (LC-MS/MS) and Application to Linseed Oil.
Douny C, et al.
Food Analytical Methods, 1-11 (2014)
Structural Dynamics of 3-Dimethylamino-2-methyl-propenal in S2 (pp*) State.
Pan S, et al.
Chinese Journal of Chemical Physics, 27(2), 149-158 (2014)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service