Skip to Content
Merck
All Photos(1)

Documents

365181

Sigma-Aldrich

N-Benzylhydroxylamine hydrochloride

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5CH2NHOH · HCl
CAS Number:
Molecular Weight:
159.61
Beilstein:
507948
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

mp

108-110 °C (lit.)

SMILES string

Cl.ONCc1ccccc1

InChI

1S/C7H9NO.ClH/c9-8-6-7-4-2-1-3-5-7;/h1-5,8-9H,6H2;1H

InChI key

YSNXOQGDHGUKCZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

N-Benzylhydroxylamine hydrochloride is a N-substituted-hydroxylamine. It participates in the ring opening of (2S,3R)-1,2-epoxy-4-penten-3-ol. Two-step synthesis of N-benzylhydroxylamine starting from dibenzylamine is reported.

Application

N-Benzylhydroxylamine hydrochloride may be used in the preparation of a precursor to hitherto unknown aminocyclopentitol derivative, hydroxy functionalised 4-exo-ethoxycarbonyl-3-oxa-2-azabicyclo[3.3.0]octane system.

Analysis Note

May contain traces of methyl orange, which can cause the color to take on a faint red hue.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

[1, 3]-Dipolar intramolecular nitrone olefin cycloaddition reaction of a sugar-derived a, ?-unsaturated ester: a new diastereo-and regioselective synthesis of an aminocyclopentitol.
Jachak SM, et al.
Tetrahedron Letters, 42(29), 4925-4928 (2001)
The Synthesis of Chiral Polyhydroxylated Pyrrolidines Using a Reverse-Cope Cyclisation.
O'Neil IA, et al.
Synlett, 10, 1408-1410 (2000)
A Large-Scale Low-Cost Preparation of N-Benzylhydroxylamine Hydrochloride.
Nguyen TB, et al.
Synthesis, 18, 3174-3176 (2009)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service