Skip to Content
Merck
All Photos(1)

Key Documents

340995

Sigma-Aldrich

Bis(2,2,2-trifluoroethyl) phosphite

technical grade, 90%

Synonym(s):

Phosphonic acid bis(2,2,2-trifluoroethyl) ester

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CF3CH2O)2P(O)H
CAS Number:
Molecular Weight:
246.04
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

grade

technical grade

Assay

90%

form

solid

refractive index

n20/D 1.332 (lit.)

bp

43-44 °C/2 mmHg (lit.)

density

1.545 g/mL at 25 °C (lit.)

functional group

fluoro

SMILES string

FC(F)(F)CO[PH](=O)OCC(F)(F)F

InChI

1S/C4H5F6O3P/c5-3(6,7)1-12-14(11)13-2-4(8,9)10/h14H,1-2H2

InChI key

NYQKUVUXMIHXEA-UHFFFAOYSA-N

Application

Bis(2,2,2-trifluoroethyl) phosphite was used in the synthesis of bis(2,2,2-trifluoroethyl phosphorochloridate.[1] It was also employed as reagent for the synthesis of mono- and diesters of phosphorous acid.[2]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

168.8 °F - closed cup

Flash Point(C)

76 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Improved synthesis of bis (2, 2, 2-trifluoroethyl) phosphorochloridate.
Bowman RA, et al.
Organic Prep. and Proc. Int., 31(2), 230-231 (1999)
Bis [2, 2, 2-trifluoroethyl] phosphite, a new reagent for synthesizing mono-and diesters of phosphorous acid.
Gibbs DE and Larsen C.
Synthesis, 5, 410-413 (1984)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service