Skip to Content
Merck
All Photos(1)

Key Documents

340294

Sigma-Aldrich

Ethyl trans-3-hexenoate

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C2H5CH=CHCH2CO2C2H5
CAS Number:
Molecular Weight:
142.20
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

liquid

refractive index

n20/D 1.426 (lit.)

bp

63-64 °C/12 mmHg (lit.)

density

0.896 g/mL at 25 °C (lit.)

functional group

ester

SMILES string

[H]\C(CC)=C(\[H])CC(=O)OCC

InChI

1S/C8H14O2/c1-3-5-6-7-8(9)10-4-2/h5-6H,3-4,7H2,1-2H3/b6-5+

InChI key

VTSFIPHRNAESED-AATRIKPKSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Ethyl trans-3-hexenoate is an ester. It was evaluated as aroma volatile in the GC effluents of cashew apple water phase phase.[1]

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Hazard Classifications

Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

138.2 °F - closed cup

Flash Point(C)

59 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Karina L Sampaio et al.
Journal of the science of food and agriculture, 91(10), 1801-1809 (2011-06-18)
There is a considerable loss of volatile compounds during the thermal concentration of cashew apple juice, damaging product quality, and as yet there is little research on the subject. Thus the purpose of this research was to identify the aroma
Yuwadee Ackarabanpojoue et al.
Journal of food science, 80(5), E998-1004 (2015-04-02)
This study aimed at investigating the effect of nitrate removal from pineapple juice by electrodialysis (ED) on selected properties of the ED-treated juice. Single-strength pineapple juice with reduced pulp content was treated by ED to reduce the nitrate concentration to

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service