Skip to Content
Merck
All Photos(1)

Key Documents

339865

Sigma-Aldrich

4-(Trifluoromethoxy)phenol

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CF3OC6H4OH
CAS Number:
Molecular Weight:
178.11
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

form

liquid

refractive index

n20/D 1.447 (lit.)

bp

92 °C/25 mmHg (lit.)

density

1.375 g/mL at 25 °C (lit.)

functional group

fluoro

SMILES string

Oc1ccc(OC(F)(F)F)cc1

InChI

1S/C7H5F3O2/c8-7(9,10)12-6-3-1-5(11)2-4-6/h1-4,11H

InChI key

WDRJNKMAZMEYOF-UHFFFAOYSA-N

Application

4-(Trifluoromethoxy)phenol was employed in the catalytic enantioselective synthesis of esters.[1]

Pictograms

Skull and crossbonesCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Aquatic Chronic 3 - Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 2

Flash Point(F)

186.8 °F - closed cup

Flash Point(C)

86 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sheryl L Wiskur et al.
Journal of the American Chemical Society, 127(17), 6176-6177 (2005-04-28)
By building on elementary principles of Brønsted acid-base chemistry, a nucleophile-catalyzed method for the asymmetric synthesis of esters from ketenes has been transformed into a much more versatile and effective Brønsted acid-catalyzed process. The product aryl esters can be converted
Kun Qian et al.
Dalton transactions (Cambridge, England : 2003), 47(36), 12759-12768 (2018-08-29)
Dielectric polymer-based nanocomposites have attracted significant attention in recent years for energy storage applications because of their potential high permittivity and breakdown strength. The coupling effect of a nanofiller/matrix interface plays a crucial role in the dielectric and electric properties

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service