Skip to Content
Merck
All Photos(1)

Key Documents

337900

Sigma-Aldrich

5-Methyl-2-thienylmethyl benzoate

technical grade, 90%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H12O2S
CAS Number:
Molecular Weight:
232.30
MDL number:

grade

technical grade

Assay

90%

SMILES string

Cc1ccc(COC(=O)c2ccccc2)s1

InChI

1S/C13H12O2S/c1-10-7-8-12(16-10)9-15-13(14)11-5-3-2-4-6-11/h2-8H,9H2,1H3

InChI key

OYCPSLZIFRQTQO-UHFFFAOYSA-N


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service