Skip to Content
Merck
All Photos(1)

Documents

333514

Sigma-Aldrich

Allyltriphenylphosphonium chloride

99%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH2=CHCH2P(Cl)(C6H5)3
CAS Number:
Molecular Weight:
338.81
Beilstein:
4169778
EC Number:
MDL number:
UNSPSC Code:
12352000

Assay

99%

SMILES string

[Cl-].C=CC[P+](c1ccccc1)(c2ccccc2)c3ccccc3

Looking for similar products? Visit Product Comparison Guide


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service