Skip to Content
Merck
All Photos(1)

Key Documents

330515

Sigma-Aldrich

(S)-(+)-2-Pentanol

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CH2CH2CH(OH)CH3
CAS Number:
Molecular Weight:
88.15
Beilstein:
1718820
MDL number:
UNSPSC Code:
12352001
PubChem Substance ID:
NACRES:
NA.22

vapor density

3 (vs air)

Assay

98%

form

liquid

optical activity

[α]25/D +13°, neat

autoignition temp.

657 °F

expl. lim.

9 %

refractive index

n20/D 1.406 (lit.)

bp

118-119 °C (lit.)

density

0.81 g/mL at 25 °C (lit.)

functional group

hydroxyl

SMILES string

CCC[C@H](C)O

InChI

1S/C5H12O/c1-3-4-5(2)6/h5-6H,3-4H2,1-2H3/t5-/m0/s1

InChI key

JYVLIDXNZAXMDK-YFKPBYRVSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

FlameExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Inhalation - Flam. Liq. 3 - STOT SE 3

Target Organs

Respiratory system

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

93.2 °F - closed cup

Flash Point(C)

34 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Xiao-Hong Chen et al.
PloS one, 9(4), e94543-e94543 (2014-04-18)
A novel carbonyl reductase (AcCR) catalyzing the asymmetric reduction of ketones to enantiopure alcohols with anti-Prelog stereoselectivity was found in Acetobacter sp. CCTCC M209061 and enriched 27.5-fold with an overall yield of 0.4% by purification. The enzyme showed a homotetrameric

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service