Skip to Content
Merck
All Photos(1)

Key Documents

32778

Sigma-Aldrich

2,4-Diaminobenzenesulfonic acid

≥98.0% (T)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H8N2O3S
CAS Number:
Molecular Weight:
188.20
Beilstein:
2694057
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

≥98.0% (T)

form

powder

mp

260-266 °C (dec.)

functional group

sulfonic acid

SMILES string

Nc1ccc(c(N)c1)S(O)(=O)=O

InChI

1S/C6H8N2O3S/c7-4-1-2-6(5(8)3-4)12(9,10)11/h1-3H,7-8H2,(H,9,10,11)

InChI key

JVMSQRAXNZPDHF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

2,4-Diaminobenzenesulfonic acid is an sulfonated aromatic amine and its aerobic biodegradation was reported.[1]

Application

2,4-Diaminobenzenesulfonic acid was used in preparation of series of sulfonated homo- and random co-polyimides.[2]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Synthesis and characterization of sulfonated homo-and co-polyimides based on 2, 4 and 2, 5-diaminobenzenesulfonic acid for proton exchange membranes.
Deligoz H, et al.
Polymers For Advanced Technologies, 19(12), 1792-1802 (2008)
A Ghaee et al.
Environmental technology, 38(6), 772-781 (2017-02-14)
Exclusion due to membrane surface charge is considered as one of the main separation mechanisms occurring in charged membranes, which can be varied through various approaches to affect membrane rejection performance. In this study, thin-film composite (TFC) polyamide (PA) membranes
[Toxicity of several derivatives of aromatic amines].
A A Silaev et al.
Meditsina truda i promyshlennaia ekologiia, (10)(10), 36-40 (1996-01-01)
Iñigo Díez-García et al.
Polymers, 11(7) (2019-07-25)
This research work was focused on the incorporation of TiO2 nanoparticles into synthesized solvent-free waterborne poly(urethane-urea) (WPUU) based on hydrophilic poly(ethylene oxide) (PU0) in order to improve both the mechanical properties and self-healing effectiveness of a polymer matrix. The incorporation
Nico C G Tan et al.
Biodegradation, 16(6), 527-537 (2005-05-04)
Ten sulfonated aromatic amines were tested for their aerobic and anaerobic biodegradability and toxicity potential in a variety of environmental inocula. Of all the compounds tested, only two aminobenzenesulfonic acid (ABS) isomers, 2- and 4-ABS, were degraded. The observed degradation

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service