Skip to Content
Merck
All Photos(1)

Key Documents

325201

Sigma-Aldrich

Ethyl 2-methylpyridine-3-carboxylate

97%

Synonym(s):

Ethyl 2-methylnicotinate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H11NO2
CAS Number:
Molecular Weight:
165.19
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

liquid

refractive index

n20/D 1.505 (lit.)

bp

126-127 °C/24 mmHg (lit.)

density

1.072 g/mL at 25 °C (lit.)

functional group

ester

SMILES string

CCOC(=O)c1cccnc1C

InChI

1S/C9H11NO2/c1-3-12-9(11)8-5-4-6-10-7(8)2/h4-6H,3H2,1-2H3

InChI key

TZORNSWZUZDUCU-UHFFFAOYSA-N

Application

Ethyl 2-methylpyridine-3-carboxylate was used in the preparation of aminoesters:
  • ethyl 2(piperidinylaminomethyl)benzoate
  • ethyl 2-(N-benzyl-N-methylaminomethyl)benzoate
  • ethyl 2-(diethylaminomethyl)benzoate

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

217.4 °F - closed cup

Flash Point(C)

103 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tandem Wolff rearrangement-?a-cyclization of tertiary amines? sequence: Synthesis of some 1H-2-benzopyran derivatives.
Leost F, et al.
Tetrahedron, 54(23), 6457-6474 (1998)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service