Skip to Content
Merck
All Photos(1)

Key Documents

309575

Sigma-Aldrich

Pentacyclo[5.4.0.02,6.03,10.05,9]undecane-8,11-dione

98%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H10O2
CAS Number:
Molecular Weight:
174.20
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

mp

240-242 °C (lit.)

functional group

ketone

SMILES string

O=C1[C@H]2[C@@H]3C[C@H]4[C@H]5[C@@H]3[C@@H]1[C@H]5C(=O)[C@@H]24

InChI

1S/C11H10O2/c12-10-6-2-1-3-5-4(2)8(10)9(5)11(13)7(3)6/h2-9H,1H2/t2-,3+,4-,5+,6-,7-,8-,9-/m1/s1

InChI key

WTUFOKOJVXNYTJ-MFAOOUEMSA-N

Application

Pentacyclo[5.4.0.02,6.03,10.05,9]undecane-8,11-dione was used in the synthesis of 8,11-dihydroxy-pentacyclo[5.4.0.02,6.03,10.05,9]undecane-8,11-lactam by reacting with aqueous sodium cyanide.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Martins FJ, et al.
Tetrahedron, 49(42) (1993)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service