Skip to Content
Merck
All Photos(1)

Documents

302201

Sigma-Aldrich

4-(Dimethylamino)-1,2,2,6,6-pentamethylpiperidine

99%

Synonym(s):

N,N,1,2,2,6,6-Heptamethyl-4-piperidinamine

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

99%

form

solid

mp

32-34 °C (lit.)

solubility

dichloromethane: soluble 25 mg/mL, clear, colorless

SMILES string

CN(C)C1CC(C)(C)N(C)C(C)(C)C1

InChI

1S/C12H26N2/c1-11(2)8-10(13(5)6)9-12(3,4)14(11)7/h10H,8-9H2,1-7H3

InChI key

VEBFUHQIQCLTJD-UHFFFAOYSA-N

Application

4-(Dimethylamino)-1,2,2,6,6-pentamethylpiperidine was used as reactant for consecutive SNAr-dequaternization reactions of heteroaromatic chlorides with tertiary amines

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

186.8 °F - closed cup

Flash Point(C)

86 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Selectivity in the Consecutive SNAr-Dequaternization Reactions of Heteroaromatic Chlorides with Tertiary Amines under High Pressure.
K. Matsumoto, et al.,
Chemistry Letters (Jpn), 8, 1283-1286 (1991)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service