Skip to Content
Merck
All Photos(1)

Documents

301337

Sigma-Aldrich

Ethyl 3,5-dinitrobenzoate

99%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(O2N)2C6H3CO2C2H5
CAS Number:
Molecular Weight:
240.17
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

99%

form

solid

mp

94-95 °C (lit.)

SMILES string

CCOC(=O)c1cc(cc(c1)[N+]([O-])=O)[N+]([O-])=O

InChI

1S/C9H8N2O6/c1-2-17-9(12)6-3-7(10(13)14)5-8(4-6)11(15)16/h3-5H,2H2,1H3

InChI key

IBQREHJPMPCXQA-UHFFFAOYSA-N

General description

Ethyl 3,5-dinitrobenzoate forms charge transfer spectra with n-alkane solutions containing hexakis(n-hexyloxy)triphenylene. Nuclear magnetic resonance spectra of ethyl 3,5-dinitrobenzoate was studied.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The mechanism of action of ethanolamine deaminase. I. Studies with isotopic hydrogen and oxygen.
B M Babior
The Journal of biological chemistry, 244(2), 449-456 (1969-01-25)
Aggregates of hexakis (n-hexyloxy) triphenylene self-assemble in dodecane solution: intercalation of (-)-menthol 3, 5-dinitrobenzoate induces formation of helical structures.
Gallivan JP and Schuster GB.
The Journal of Organic Chemistry, 60(8), 2423-2429 (1995)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service