All Photos(1)
About This Item
Linear Formula:
(C2H5)2SiHCH3
CAS Number:
Molecular Weight:
102.25
Beilstein:
1730936
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Assay
95%
form
liquid
refractive index
n20/D 1.398 (lit.)
bp
78 °C (lit.)
density
0.705 g/mL at 25 °C (lit.)
SMILES string
CC[SiH](C)CC
InChI
1S/C5H14Si/c1-4-6(3)5-2/h6H,4-5H2,1-3H3
InChI key
UUTZEAQZDHSEAL-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
accessory
Product No.
Description
Pricing
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2 - Flam. Liq. 2 - Skin Irrit. 2 - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
3 - Flammable liquids
WGK
WGK 1
Flash Point(F)
-9.4 °F - closed cup
Flash Point(C)
-23 °C - closed cup
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service