Skip to Content
Merck
All Photos(1)

Key Documents

273104

Sigma-Aldrich

3,5-Di-tert-butylaniline

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[(CH3)3C]2C6H3NH2
CAS Number:
Molecular Weight:
205.34
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

mp

54-57 °C (lit.)

SMILES string

CC(C)(C)c1cc(N)cc(c1)C(C)(C)C

InChI

1S/C14H23N/c1-13(2,3)10-7-11(14(4,5)6)9-12(15)8-10/h7-9H,15H2,1-6H3

InChI key

MJKNHXCPGXUEDO-UHFFFAOYSA-N

Application

3,5-Di-tert-butylaniline was used in the synthesis of lipophilic wedges by reacting with 2-amino-6-chloropyrimidin-4-ol in a refluxing mixture of acetic and hydrochloric acid[1].

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lakshya Daukiya et al.
Nanoscale, 13(5), 2972-2981 (2021-01-29)
Covalent functionalization is one of the most efficient ways to tune the properties of layered materials in a highly controlled manner. However, molecular chemisorption on semiconducting transition metal dichalcogenides remains a delicate task due to the inertness of their surface.
Thi Mien Trung Huynh et al.
Nanoscale, 9(1), 362-368 (2016-12-08)
Highly oriented pyrolytic graphite (HOPG) can be covalently grafted with aryl radicals generated via the electrochemical reduction of 3,5-bis-tert-butyl-diazonium cations (3,5-TBD). The structure of the grafted layer and its stability under electrochemical conditions were assessed with electrochemical scanning tunneling microscopy
JANUS WEDGES: a new approach towards nucleobase-pair recognition.
Chemical Communications (Cambridge, England), 21, 2443-2444 (1996)
Gina Ambrosio et al.
Scientific reports, 10(1), 4114-4114 (2020-03-07)
Chemical reaction with diazonium molecules has revealed to be a powerful method for the surface chemical modification of graphite, carbon nanotubes and recently also of graphene. Graphene electronic structure modification using diazonium molecules is strongly influenced by graphene growth and
Ji Han Kim et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 25(38), 9060-9070 (2019-04-16)
Organic light-emitting diodes are currently under research to achieve high efficiency and long life by using thermally activated delayed fluorescence (TADF) materials. In particular, many studies have focused on ensuring high efficiency in fluorescent devices by introducing TADF materials. Herein

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service