Skip to Content
Merck
All Photos(1)

Documents

272434

Sigma-Aldrich

Bicyclo[3.3.1]nonane-3,7-dione

98%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H12O2
CAS Number:
Molecular Weight:
152.19
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

form

powder

SMILES string

O=C1C[C@H]2C[C@@H](C1)CC(=O)C2

InChI

1S/C9H12O2/c10-8-2-6-1-7(4-8)5-9(11)3-6/h6-7H,1-5H2/t6-,7+

InChI key

KIKCULSOJJAIEB-KNVOCYPGSA-N

General description

The high stability and formation of the supramolecular structures of the syn and anti isomers of the dioxime of bicyclo[3.3.1]nonane-3,7-dione were studied.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

S Gailani et al.
Journal of medicine, 8(6), 403-414 (1977-01-01)
A 76-year-old man with plasma cell leukemia was treated with L-phenylalanine mustard and prednisone (CLGB 7461). There was good partial remission of the plasma cell disease characterized by disappearance of the plasma cells in the peripheral blood, reduction of plasma

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service