Skip to Content
Merck
All Photos(1)

Documents

264415

Sigma-Aldrich

2,4-Difluorobenzyl bromide

98%

Synonym(s):

α-Bromo-2,4-difluorotoluene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
F2C6H3CH2Br
CAS Number:
Molecular Weight:
207.02
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

form

liquid

refractive index

n20/D 1.525 (lit.)

density

1.613 g/mL at 25 °C (lit.)

SMILES string

Fc1ccc(CBr)c(F)c1

InChI

1S/C7H5BrF2/c8-4-5-1-2-6(9)3-7(5)10/h1-3H,4H2

InChI key

IBLMYGXJKQIGSN-UHFFFAOYSA-N

Application

2,4-Difluorobenzyl bromide has been used in the preparation of:
  • novel 1,2,4-triazolium derivatives
  • 1,5-biaryl pyrrole EP1 receptor antagonists

Signal Word

Danger

Hazard Statements

Hazard Classifications

Flam. Liq. 3 - Resp. Sens. 1 - Skin Corr. 1B - Skin Sens. 1

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

104.0 °F - closed cup

Flash Point(C)

40 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Adrian Hall et al.
Bioorganic & medicinal chemistry letters, 16(14), 3657-3662 (2006-05-16)
The preliminary SAR of a series of novel 1,5-biaryl pyrrole EP1 receptor antagonists derived from compound 1 is described. Replacement of the benzyl group of 1 with isosteric groups was investigated. The most effective replacement was found to be the
Yan Luo et al.
Archiv der Pharmazie, 342(7), 386-393 (2009-06-23)
A series of novel 1,2,4-triazolium derivatives was synthesized starting from commercially available 1H-1,2,4-triazole, 2,4-dichlorobenzyl chloride, or 2,4-difluorobenzyl bromide. Their antibacterial and antifungal activities were evaluated against Staphylococcus aureus ATCC 29213, Escherichia coli ATCC 25922, Bacillus proteus, Bacillus subtilis, Pseudomonas aeruginosa

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service