Skip to Content
Merck
All Photos(2)

Key Documents

255998

Sigma-Aldrich

Antimony(V) oxide

99.99% trace metals basis

Synonym(s):

Antimonic oxide, Diantimony pentaoxide, Diantimony pentoxide, Antimony pentoxide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Sb2O5
CAS Number:
Molecular Weight:
323.52
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
NACRES:
NA.22

Assay

99.99% trace metals basis

form

solid

reaction suitability

core: antimony
reagent type: catalyst

mp

380 °C (dec.) (lit.)

density

3.78 g/mL at 25 °C (lit.)

SMILES string

O=[Sb](=O)O[Sb](=O)=O

InChI

1S/5O.2Sb

InChI key

LJCFOYOSGPHIOO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Use as a flame retardant in ABS and other plastics, a flocculant in the production of titanium dioxide and is sometimes used in the production of glass, paint and adhesives; and also used as an ion-exchange resin and as a polymerization and oxidation catalyst.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

S Miyata et al.
Clinica chimica acta; international journal of clinical chemistry, 106(3), 235-242 (1980-10-09)
A rapid and accurate method is presented for the determination of manganese in biological samples, using neutron activation analysis. Biological samples were irradiated at 5000 kW for 30 min. The samples were ashed on a hot plate with 14 mol/l
V Potkonjak et al.
International archives of occupational and environmental health, 51(3), 199-207 (1983-01-01)
The occupational exposure, clinical findings, lung function and radiological characteristics of 51 workers in an antimony smelting plant are presented. The workers were exposed to the airbone dust containing a high concentration of antimony trioxide (up to 88%) and antimony

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service