Skip to Content
Merck
All Photos(1)

Key Documents

254452

Sigma-Aldrich

Chlorotripropylsilane

98%

Synonym(s):

Tripropylchlorosilane

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3CH2CH2)3SiCl
CAS Number:
Molecular Weight:
192.80
Beilstein:
1737489
EC Number:
MDL number:
UNSPSC Code:
12352001
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

refractive index

n20/D 1.44 (lit.)

bp

202 °C (lit.)

density

0.882 g/mL at 25 °C (lit.)

SMILES string

CCC[Si](Cl)(CCC)CCC

InChI

1S/C9H21ClSi/c1-4-7-11(10,8-5-2)9-6-3/h4-9H2,1-3H3

InChI key

ACTAPAGNZPZLEF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Skin Corr. 1A - STOT SE 3

Target Organs

Respiratory system

Supplementary Hazards

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

145.4 °F - closed cup

Flash Point(C)

63 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service