Skip to Content
Merck
All Photos(1)

Key Documents

247987

Sigma-Aldrich

1-Bromo-2,4,6-trifluorobenzene

99%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
BrC6H2F3
CAS Number:
Molecular Weight:
210.98
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

99%

form

liquid

refractive index

n20/D 1.485 (lit.)

bp

140.5 °C (lit.)

mp

3.5 °C (lit.)

density

1.79 g/mL at 25 °C (lit.)

SMILES string

Fc1cc(F)c(Br)c(F)c1

InChI

1S/C6H2BrF3/c7-6-4(9)1-3(8)2-5(6)10/h1-2H

InChI key

PZBSPSOGEVCRQI-UHFFFAOYSA-N

General description

1-Bromo-2,4,6-trifluorobenzene exhibits packing polymorphism in two nonidentical solid-state phases[1]. 1-Bromo-2,4,6-trifluorobenzene participates in Pd-catalyzed oxidative Heck reaction with allyl ester[2].

Pictograms

FlameExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

136.4 °F - closed cup

Flash Point(C)

58 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Zejiang Li et al.
Organic letters, 14(1), 74-77 (2011-12-14)
An efficient Pd(II)-catalyzed direct olefination of perfluoroarenes with allyl esters is demonstrated. Under the typical conditions, the coupling reaction of fluorinated-arenes with allylic esters proceeded via a β-H elimination rather than a β-OAc elimination to give the corresponding γ-substituted allylic
Harry G Brittain
Journal of pharmaceutical sciences, 101(2), 464-484 (2011-10-13)
Papers and patents that deal with polymorphism (crystal systems for which a substance can exist in structures defined by different unit cells and where each of the forms has the same elemental composition) and solvatomorphism (systems where the crystal structures

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service