Skip to Content
Merck
All Photos(1)

Key Documents

247308

Sigma-Aldrich

(S)-(+)-2-Methylbutyronitrile

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C2H5CH(CH3)CN
CAS Number:
Molecular Weight:
83.13
MDL number:
UNSPSC Code:
12352001
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

optical activity

[α]20/D +31°, neat

refractive index

n20/D 1.39 (lit.)

bp

125-126 °C (lit.)

density

0.786 g/mL at 25 °C (lit.)

functional group

nitrile

storage temp.

2-8°C

SMILES string

CC[C@H](C)C#N

InChI

1S/C5H9N/c1-3-5(2)4-6/h5H,3H2,1-2H3/t5-/m0/s1

InChI key

RCEJCSULJQNRQQ-YFKPBYRVSA-N

Application

Chiral building block

Pictograms

FlameExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

75.2 °F - closed cup

Flash Point(C)

24 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service