Skip to Content
Merck
All Photos(1)

Key Documents

233048

Sigma-Aldrich

4-(2-Furyl)-3-buten-2-one, predominantly cis

95%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H8O2
CAS Number:
Molecular Weight:
136.15
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

95%

form

solid

refractive index

n20/D 1.565 (lit.)

mp

34-41 °C (lit.)

SMILES string

CC(=O)\C=C/c1ccco1

InChI

1S/C8H8O2/c1-7(9)4-5-8-3-2-6-10-8/h2-6H,1H3/b5-4-

InChI key

GBKGJMYPQZODMI-PLNGDYQASA-N

General description

4-(2-Furyl)-3-buten-2-one participates in base catalyzed aldol condensation of furfurals with acetone or propanal. It also participates in aldol-condensation and hydrogenation reactions of synthetic bio-oils at 100°C and 400 psi in a biphasic system of decalin/water.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

219.2 °F

Flash Point(C)

104 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Condensation/hydrogenation of biomass-derived oxygenates in water/oil emulsions stabilized by nanohybrid catalysts.
Zapata PA, et al.
Topics in Catalysis, 55(1-2), 38-52 (2012)
Liquid phase aldol condensation reactions with MgO-ZrO2 and shape-selective nitrogen-substituted NaY.
Shen W, et al.
Applied Catalysis A: General, 392(1), 57-68 (2011)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service