Skip to Content
Merck
All Photos(1)

Documents

226327

Sigma-Aldrich

Ethyl O-(2-mesitylenesulfonyl)acethydroxamate

98%

Synonym(s):

O-(2-Mesitylenesulfonyl)acethydroxamic acid ethyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
2,4,6-(CH3)3C6H2SO2ON=C(CH3)OC2H5
CAS Number:
Molecular Weight:
285.36
Beilstein:
2144156
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

mp

54-56 °C (lit.)

storage temp.

2-8°C

SMILES string

CCO\C(C)=N\OS(=O)(=O)c1c(C)cc(C)cc1C

InChI

1S/C13H19NO4S/c1-6-17-12(5)14-18-19(15,16)13-10(3)7-9(2)8-11(13)4/h7-8H,6H2,1-5H3/b14-12+

InChI key

KQCBSWBQAXTILK-WYMLVPIESA-N

Looking for similar products? Visit Product Comparison Guide

Application

EthylO-(2-Mesitylenesulfonyl)acethydroxamic acid ethyl ester may be used in chemical synthesis studies.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service