Skip to Content
Merck
All Photos(1)

Key Documents

225436

Sigma-Aldrich

2,4-Dichloro-6-nitrophenol

≥95%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Cl2C6H2(NO2)OH
CAS Number:
Molecular Weight:
208.00
Beilstein:
2050081
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

≥95%

form

crystals

contains

20% water

mp

118-120 °C (lit.)

SMILES string

Oc1c(Cl)cc(Cl)cc1[N+]([O-])=O

InChI

1S/C6H3Cl2NO3/c7-3-1-4(8)6(10)5(2-3)9(11)12/h1-2,10H

InChI key

LYPMXMBQPXPNIQ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

The FT-Raman and FTIR spectra of 2,4-dichloro-6-nitrophenol (2,4-DC6NP) was studied.

Application

2,4-Dichloro-6-nitrophenol, a specific inhibitor of sulfotransferases, was used to reduce the neosynthesis of [3H] pregnenolone sulfate (Δ5PS) and [3H]dehydroepiandrosterone sulfate (DHEAS).

Physical form

Moist solid

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

N Sundaraganesan et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 65(5), 1053-1062 (2006-05-24)
The FTIR and FT-Raman spectra of 2,4-dichloro-6-nitrophenol (2,4-DC6NP) has been recorded in the region 4000-400 cm(-1) and 3500-100 cm(-1), respectively. The optimized geometry, frequency and intensity of the vibrational bands of (2,4-DC6NP) were obtained by the ab initio and DFT
D Beaujean et al.
Journal of neurochemistry, 72(2), 848-857 (1999-02-04)
Biosynthesis of the neuroactive steroids pregnenolone sulfate (delta5PS) and dehydroepiandrosterone sulfate (DHEAS) is catalyzed by the enzyme hydroxysteroid sulfotransferase (HST), which transfers the sulfonate moiety from 3'-phosphoadenosine 5'-phosphosulfate (PAPS) on the 3-hydroxy site of steroids. Although high concentrations of delta5PS
Tilman Pfeffer et al.
The Journal of pharmacology and experimental therapeutics, 375(3), 478-487 (2020-10-07)
The lysyl hydroxylases (procollagen-lysine 5-dioxygenases) PLOD1, PLOD2, and PLOD3 have been proposed as pathogenic mediators of stunted lung development in bronchopulmonary dysplasia (BPD), a common complication of preterm birth. In affected infants, pulmonary oxygen toxicity stunts lung development. Mice lacking

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service