Skip to Content
Merck
All Photos(1)

Key Documents

213020

Sigma-Aldrich

Malachite Green Carbinol hydrochloride

Dye content 85 %

Synonym(s):

Basic Green 4, Malachite Green hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C23H26N2O · HCl
CAS Number:
Molecular Weight:
382.93
Colour Index Number:
42000
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:

composition

Dye content, 85%

λmax

425 nm
615 nm (2nd)

SMILES string

Cl[H].CN(C)c1ccc(cc1)C(O)(c2ccccc2)c3ccc(cc3)N(C)C

InChI

1S/C23H26N2O.ClH/c1-24(2)21-14-10-19(11-15-21)23(26,18-8-6-5-7-9-18)20-12-16-22(17-13-20)25(3)4;/h5-17,26H,1-4H3;1H

InChI key

MJRWALOBODKCCZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Features and Benefits

In solution, the hydrochloride loses water rapidly to give the chloride salt that corresponds to the oxalate (cat. no. 861219).

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Yann Desfougères et al.
Methods in molecular biology (Clifton, N.J.), 2091, 59-71 (2019-11-28)
The yeast Saccharomyces cerevisiae has given us much information on the metabolism and function of inositol polyphosphates and inorganic polyphosphate. To expand our knowledge of the metabolic as well as functional connections between inositol polyphosphates and inorganic polyphosphate, we have

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service