Skip to Content
Merck
All Photos(3)

Key Documents

19979

Sigma-Aldrich

Microreactor Explorer Kit

volume 0.85 mL , Microreactor, AC/DC input 120 V / 240 V AC

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200

feature

CE compliant

flow rate measuring range

0.2-15 mL/min

temperature measuring range

-70-150 °C

parameter

6.5 bar max. pressure

volume

0.85 mL , Microreactor
25 mL , Residence time unit

General description

All-in-one solution for executing liquid/liquid reactions continuously in a microstructured reactor. This method has significant potential for process performance improvement compared to the conventional mode. The metal-free construction enables safe handling of aggressive compounds under permanent pressure and temperature supervision. Automatic shutdown function permits unattended operation. Adjust the temperature by positioning the reactor and the residence time unit in your bath which should be on a magnetic stirrer (bath and stirrer not included). Resident volumes are adjustable to individual demands.
Fields of application: General process development, high exothermic/hazardous reactions, handling of products that are instable under reaction conditions and product profile improvement (side product suppression).
Contents:
  • Glass microreactor including clamp, 1
  • Rotary piston pumps, 2
  • Thermo sensors, 2
  • Pressure sensors, 2
  • Control unit, 1
  • Residence time unit (I.D. 1/8 in. tube, PTFE)
  • Fittings, spare parts, case

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service