Skip to Content
Merck
All Photos(1)

Key Documents

196843

Sigma-Aldrich

1,2,4-Trifluorobenzene

98%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H3F3
CAS Number:
Molecular Weight:
132.08
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

form

liquid

refractive index

n20/D 1.423 (lit.)

bp

88 °C/759 mmHg (lit.)

density

1.264 g/mL at 25 °C (lit.)

functional group

fluoro

SMILES string

Fc1ccc(F)c(F)c1

InChI

1S/C6H3F3/c7-4-1-2-5(8)6(9)3-4/h1-3H

InChI key

PEBWOGPSYUIOBP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Infrared absorption spectrum of liquid 1,2,4-trifluorobenzene has been investigated using LiF, NaCl, KBr and KRS-5 prisms[1]. The microwave spectrum of 1,2,4-trifluorobenzene has been investigated at dry ice temperature in the frequency range of 8 to 12.4GHz[2].

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

39.2 °F - closed cup

Flash Point(C)

4 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Vibrational Spectra of Fluorinated Aromatics. VII. 1, 2, 4-Trifluorobenzene.
Ferguson EE, et al.
J. Chem. Phys. , 21(10), 1727-1730 (2004)
Microwave Spectrum, Dipole Moment and Centrifugal Distortion Constants of 1, 2, 4-Trifluorobenzene.
Doraiswamy S and Sharma SD.
Journal of the Physical Society of Japan, 44(2), 598-601 (1978)
Haiyang Zhang et al.
Journal of chromatography. A, 1440, 66-73 (2016-02-27)
A hybrid fluorous monolithic column was simply prepared via photo-initiated free radical polymerization of an acrylopropyl polyhedral oligomeric silsesquioxane (acryl-POSS) and a perfluorous monomer (2,2,3,3,4,4,5,5,6,6,7,7-dodecafluoroheptyl acrylate) in UV-transparent fused-silica capillaries within 5min. The physical characterization of hybrid fluorous monolith, including
Julian Renpenning et al.
Rapid communications in mass spectrometry : RCM, 31(13), 1095-1102 (2017-04-05)
The conventional high-temperature conversion (HTC) approach towards hydrogen compound-specific isotope analysis (CSIA) of halogen-bearing (F, Cl, Br, I) organics suffers from incomplete H The performance of the modified gas chromatography/chromium-based high-temperature conversion (GC-Cr/HTC) system was monitored and optimized using an

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service