Skip to Content
Merck
All Photos(1)

Documents

196770

Sigma-Aldrich

2,3,4,6-Tetrafluoronitrobenzene

≥97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HC6F4NO2
CAS Number:
Molecular Weight:
195.07
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

≥97%

form

liquid

refractive index

n20/D 1.464 (lit.)

density

1.511 g/mL at 25 °C (lit.)

SMILES string

[O-][N+](=O)c1c(F)cc(F)c(F)c1F

InChI

1S/C6HF4NO2/c7-2-1-3(8)6(11(12)13)5(10)4(2)9/h1H

InChI key

FDLCUAUNAWWSBX-UHFFFAOYSA-N

General description

Molluscicidal activity of 2,3,4,6-tetrafluoronitrobenzene against Biomphalaria glabrata has been investigated. Palladium-catalyzed cross-coupling reactions of 2,3,4,6-tetrafluoronitrobenzene with alkyne derivatives has been studied. Reaction of 2,3,4,6-tetrafluoronitrobenzene with 7-methyl-1,5,7-triazabicyclo[4.4.0]dec-5-ene in the presence of water molecules in acetonitrile has been investigated.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

159.8 °F - closed cup

Flash Point(C)

71 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Pd-catalyzed sp2-sp cross-coupling reactions involving C-F bond activation in highly fluorinated nitrobenzene systems.
Cargill MR, et al.
Tetrahedron, 69(2), 512-516 (2013)
Reactions of fluoronitrobenzenes with MTBD strong base in acetonitrile in the presence of water molecules.
Gierczyk B, et al.
Journal of Molecular Structure, 655(2), 259-267 (2003)
Effects of structural rearrangement on the molluscicidal activity of certain fluorinated aromatic compounds.
J Duncan et al.
Bulletin of the World Health Organization, 42(5), 820-825 (1970-01-01)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service