Skip to Content
Merck
All Photos(1)

Key Documents

195723

Sigma-Aldrich

3,4-Dihydro-2,2-dimethyl-4-oxo-2H-pyran-6-carboxylic acid

98%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H10O4
CAS Number:
Molecular Weight:
170.16
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

mp

170 °C (dec.) (lit.)

functional group

carboxylic acid
ether
ketone

SMILES string

CC1(C)CC(=O)C=C(O1)C(O)=O

InChI

1S/C8H10O4/c1-8(2)4-5(9)3-6(12-8)7(10)11/h3H,4H2,1-2H3,(H,10,11)

InChI key

ANKQOBXQEHGUJT-UHFFFAOYSA-N

General description

Butyl ester of 3,4-dihydro-2,2-dimethyl-4-oxo-2H-pyran-6-carboxylic acid is commonly used as mosquito repellent.

Application

3,4-Dihydro-2,2-dimethyl-4-oxo-2H-pyran-6-carboxylic acid was used as photoprotective reagent during photolysis of the Salmonella sialidase.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J A Thomson et al.
Cancer research, 37(9), 3266-3273 (1977-09-01)
Protein phosphokinases were isolated from the nuclei of normal and fetal liver and neoplastic tissues. Chromatography on phosphocellulose columns resolved the normal and fetal liver kinases into five reproducible fractions. Each of the fractions differed in optimal divalent cation and
T G Warner et al.
The Biochemical journal, 285 ( Pt 3), 957-964 (1992-08-01)
The sialidase from Salmonella typhimurium LT2 was characterized by using photoaffinity-labelling techniques. The well-known sialidase inhibitor 5-acetamido-2,6-anhydro-3,5-dideoxy-D-glycero-D-galacto-non- 2-enonic acid (Neu5Ac2en) was modified to contain an amino group at C-9, which permitted the incorporation of 4-azidosalicylic acid in amide linkage at

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service