Skip to Content
Merck
All Photos(1)

Documents

195715

Sigma-Aldrich

Isobutyl benzoate

99%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5CO2CH2CH(CH3)2
CAS Number:
Molecular Weight:
178.23
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

99%

refractive index

n20/D 1.495 (lit.)

bp

241-242 °C (lit.)

density

0.996 g/mL at 25 °C (lit.)

SMILES string

CC(C)COC(=O)c1ccccc1

InChI

1S/C11H14O2/c1-9(2)8-13-11(12)10-6-4-3-5-7-10/h3-7,9H,8H2,1-2H3

InChI key

KYZHGEFMXZOSJN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

204.8 °F - closed cup

Flash Point(C)

96.00 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

[Rationale for maximum allowable concentration of isobutyl benzoate in atmospheric air of populated areas].
M V Murkova et al.
Gigiena i sanitariia, (2)(2), 15-16 (1994-02-01)
Corine Tourné-Péteilh et al.
Physical chemistry chemical physics : PCCP, 13(34), 15523-15529 (2011-07-30)
Interfacial tension, electrical conductivity, NMR self-diffusion and DLS experiments have been used to investigate the self-aggregation in water of ionic liquids associating an ibuprofenate anion and 1-alkyl-3-methylimidazolium [C(n)MIm](+) (n = 4, 6, 8) cations. Despite the short alkyl chain on

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service