Skip to Content
Merck
All Photos(1)

Key Documents

193208

Sigma-Aldrich

Methyl 2-benzylmalonamate

96%

Synonym(s):

Benzylmalonic acid methyl ester monoamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H13NO3
CAS Number:
Molecular Weight:
207.23
MDL number:

Assay

96%

SMILES string

COC(=O)C(Cc1ccccc1)C(N)=O

InChI

1S/C11H13NO3/c1-15-11(14)9(10(12)13)7-8-5-3-2-4-6-8/h2-6,9H,7H2,1H3,(H2,12,13)

InChI key

MBGQTVLDUWAMMN-UHFFFAOYSA-N


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service