Skip to Content
Merck
All Photos(1)

Documents

17241

Sigma-Aldrich

(R)-(−)-2-[(RS)-1-Ethoxyethoxy]-1-propanol

≥95.0% (sum of enantiomers, GC)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H16O3
CAS Number:
Molecular Weight:
148.20
Beilstein:
7566593
MDL number:
UNSPSC Code:
12352001
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

≥95.0% (sum of enantiomers, GC)

optical activity

[α]20/D −18.5±1°, neat

refractive index

n20/D 1.420

density

0.951 g/mL at 20 °C (lit.)

SMILES string

CCOC(C)O[C@H](C)CO

InChI

1S/C7H16O3/c1-4-9-7(3)10-6(2)5-8/h6-8H,4-5H2,1-3H3/t6-,7?/m1/s1

InChI key

VLECUSKDVHMJKF-ULUSZKPHSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service