Skip to Content
Merck
All Photos(2)

Documents

158259

Sigma-Aldrich

O-Methyl-L-tyrosine

98%

Synonym(s):

4-Methoxy-L-phenylalanine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3OC6H4CH2CH(NH2)CO2H
CAS Number:
Molecular Weight:
195.22
Beilstein:
2212726
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

optical activity

[α]25/D −7°, c = 0.5 in 1 M HCl

reaction suitability

reaction type: solution phase peptide synthesis

mp

259-261 °C (dec.) (lit.)

application(s)

peptide synthesis

storage temp.

2-8°C

SMILES string

COc1ccc(C[C@H](N)C(O)=O)cc1

InChI

1S/C10H13NO3/c1-14-8-4-2-7(3-5-8)6-9(11)10(12)13/h2-5,9H,6,11H2,1H3,(H,12,13)/t9-/m0/s1

InChI key

GEYBMYRBIABFTA-VIFPVBQESA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Debbie Dewaele et al.
Rapid communications in mass spectrometry : RCM, 30(6), 719-730 (2016-02-13)
Melphalan is a frequently used chemotherapeutical agent for the treatment of myeloma, breast cancer, ovarian cancer and sarcoma of soft tissue. A good knowledge of the reactivity of the drug toward the different amino acids, e.g. covalent adduct formation, is
Ma Diarey Tianero et al.
Nature microbiology, 4(7), 1149-1159 (2019-04-03)
Marine sponges often house small-molecule-producing symbionts extracellularly in their mesohyl, providing the host with a means of chemical defence against predation and microbial infection. Here, we report an intriguing case of chemically mediated symbiosis between the renieramycin-containing sponge Haliclona sp.
C Lee et al.
Biochemistry and molecular biology international, 31(5), 797-805 (1993-12-01)
An enzyme fraction with catalytic activities for the biosynthesis of the pipecolic acid containing cyclopeptolide SDZ 90-215 was partially purified and characterized from the genus Septoria. The crude cell homogenate was subjected to polyethyleneimine precipitation, ammonium sulfate precipitation and FPLC
S E Shoelson et al.
The Journal of biological chemistry, 264(14), 7831-7836 (1989-05-15)
The receptors for insulin and epidermal growth factor undergo tyrosine autophosphorylation in response to ligand stimulation, while pp60v-src is an unregulated tyrosine kinase. In this report we show that each of the kinases phosphorylates an exogenous peptide that corresponds to
Jeffrey K Takimoto et al.
ACS chemical biology, 6(7), 733-743 (2011-05-07)
Unnatural amino acids (Uaas) can be translationally incorporated into proteins in vivo using evolved tRNA/aminoacyl-tRNA synthetase (RS) pairs, affording chemistries inaccessible when restricted to the 20 natural amino acids. To date, most evolved RSs aminoacylate Uaas chemically similar to the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service