Skip to Content
Merck
All Photos(1)

Key Documents

148253

Sigma-Aldrich

Iminodiacetonitrile

technical grade

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HN(CH2CN)2
CAS Number:
Molecular Weight:
95.10
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

grade

technical grade

mp

69-71 °C (lit.)

SMILES string

N#CCNCC#N

Application

Iminodiacetonitrile was used in the synthesis of iminodiacetic acid[1]. It was also used to study the interference of nitriles in cyanide determination[2].

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

R Rubio et al.
The Analyst, 115(7), 959-963 (1990-07-01)
The interference of nitriles and isonitriles in cyanide determination was studied using the pyridine-barbituric acid direct spectrophotometric method or the same method after distillation of the sample, a microdiffusion-potentiometric (ion-selective electrode) also being used. Phenylacetonitrile, benzonitrile, acrylonitrile, iminodiacetonitrile and acetonitrile
Zhi-Qiang Liu et al.
Biotechnology progress, 27(3), 698-705 (2011-05-14)
Iminodiacetic acid (IDA) has been widely used as an important intermediate in the fine chemical industry. In this study, a novel synthesis route of IDA from iminodiacetonitrile by whole microorganisms was investigated. A strain with the capability of producing nitrilase

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service