Skip to Content
Merck
All Photos(1)

Key Documents

119903

Sigma-Aldrich

6,7-Dihydroxy-4-methylcoumarin

98%

Synonym(s):

4-Methylesculetin

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H8O4
CAS Number:
Molecular Weight:
192.17
Beilstein:
162550
EC Number:
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:

Assay

98%

mp

274-276 °C (lit.)

λmax

348 nm

SMILES string

CC1=CC(=O)Oc2cc(O)c(O)cc12

Looking for similar products? Visit Product Comparison Guide

Application

Can be used as a fluorescent sensor to monitor the consumption of a boronic acid in a Suzuki cross-coupling reaction. Fluorescence is easily detectable to the naked eye using a standard hand-held long-wave UV lamp (365 nm).

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Timothy E Barder et al.
Organic letters, 9(1), 137-139 (2006-12-29)
[reaction: see text] Although boronic acids are widely used in metal-catalyzed reactions, it is difficult to assay their consumption. As such, we developed a reversible fluorescent sensor that is activated upon binding a boronic acid. The sensor can be used

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service