Skip to Content
Merck
All Photos(1)

Documents

115762

Sigma-Aldrich

2,2,6,6-Tetramethyl-4-piperidone hydrochloride

98%

Synonym(s):

2,2,6,6-Tetramethyl-4-piperidinone hydrochloride, 2,2,6,6-Tetramethyl-4-piperidone hydrochloride, 2,2,6,6-Tetramethyl-4-piperidonium chloride, Triacetonamine hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H17NO · HCl
CAS Number:
Molecular Weight:
191.70
Beilstein:
4154554
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

form

solid

mp

198 °C (dec.) (lit.)

SMILES string

Cl[H].CC1(C)CC(=O)CC(C)(C)N1

InChI

1S/C9H17NO.ClH/c1-8(2)5-7(11)6-9(3,4)10-8;/h10H,5-6H2,1-4H3;1H

InChI key

ZXNWYMNKYXUZGM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

2,2,6,6-Tetramethyl-4-piperidone hydrochloride has been used as a reagent injected in the micromixer using a Rheodyne injection valve for introducing discrete sample pulses into the miniaturised-SYNthesis and Total Analysis System (πSYNTAS).

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Keita Saito et al.
Free radical biology & medicine, 160, 596-603 (2020-09-07)
More detailed investigations on the in vivo redox status are needed to elucidate the mechanisms contributing to damage caused by ionizing radiation. In the present study, the in vivo redox status of mice was examined using in vivo electron spin
S Dzwigaj et al.
Free radical research, 23(2), 103-115 (1995-08-01)
The production of singlet oxygen by H2O2 disproportionation and via the oxidation of H2O2 by NaOCl in a neutral medium was monitored by spin trapping with 2,2,6,6 tetramethyl-4-piperidone (TMPone). The singlet oxygen formed in both reactions oxidized 2,2,6,6 tetramethyl-4-piperidone to
K Yamamoto et al.
Biochemical pharmacology, 42(5), 1087-1092 (1991-08-08)
Photoexcited hematoporphyrin (Hp) induces the uncoupling of oxidative phosphorylation of mitochondria. The uncoupling was inhibited by pre-incubation of mitochondria with a fluorescent SH reagent, eosin-5-maleimide, which has been shown to react specifically with an essential SH group of the Pi/H+
Shaohong Ren et al.
Zhong yao cai = Zhongyaocai = Journal of Chinese medicinal materials, 27(12), 917-918 (2005-04-06)
To study the alkaloids from Armeniaca mume Sieb. The alkaloids were extracted with chloroform from 80% alcoholic extract and concentrated. Then they were transferred into aqueous solution and absorbed by macroporous cation exchange resin. The concentrated total eluent was extracted
K Takeshita et al.
Antioxidants & redox signaling, 2(2), 355-362 (2001-03-07)
The conversion of singlet oxygen ((1)O2) to hydroxyl radical (*OH) during photosensitization of uroporphyrin (UP) in the presence of NADPH was examined by a spin-trapping technique with 5,5-dimethyl-1-pyrroline-N-oxide (DMPO). Significant electron spin resonance (ESR) signals of DMPO-OH adduct were observed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service