Skip to Content
Merck
All Photos(1)

Documents

110515

Sigma-Aldrich

cis-1,4-Dimethylcyclohexane

99%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H10(CH3)2
CAS Number:
Molecular Weight:
112.21
Beilstein:
1900345
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

vapor pressure

34.2 mmHg ( 37.7 °C)

Assay

99%

autoignition temp.

579 °F

refractive index

n20/D 1.429 (lit.)

bp

124-125 °C (lit.)

mp

−88-−87 °C (lit.)

density

0.783 g/mL at 25 °C (lit.)

SMILES string

C[C@H]1CC[C@@H](C)CC1

InChI

1S/C8H16/c1-7-3-5-8(2)6-4-7/h7-8H,3-6H2,1-2H3/t7-,8+

InChI key

QRMPKOFEUHIBNM-OCAPTIKFSA-N

Looking for similar products? Visit Product Comparison Guide

Application

cis-1,4-Dimethylcyclohexane has been used to study the absorption of ultrasonic waves that can be measured by a reverberation technique.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Chronic 2 - Asp. Tox. 1 - Flam. Liq. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

42.8 °F - closed cup

Flash Point(C)

6 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service