ADVH14DBDDBF
AChemBlock
benzyl 3-methylideneazetidine-1-carboxylate
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Iupac Name
benzyl 3-methyleneazetidine-1-carboxylate
Inchi Code
1S/C12H13NO2/c1-10-7-13(8-10)12(14)15-9-11-5-3-2-4-6-11/h2-6H,1,7-9H2
InChI key
HPAXEPGOVIEEQX-UHFFFAOYSA-N
storage temp.
Store at 0-8 C
Purity
0.97
Country of Origin
CN
Physical Form
light yellow liquid
Product Link
https://www.achemblock.com/p44805-benzyl-3-methylideneazetidine-1-carboxylate.html
Synonym
benzyl 3-methyleneazetidine-1-carboxylate
Signal Word
Warning
Hazard Statements
Precautionary statements
MSDS
https://www.achemblock.com/p44805-benzyl-3-methylideneazetidine-1-carboxylate.html
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.