814479
TOYOPEARL® Phenyl-650C Bulk Media
phase phenyl, bottle of 100 mL, 100 μm particle size
Synonym(s):
TOYOPEARL® Phenyl-650 Bulk Media
About This Item
product line
TOYOPEARL®
form
slurry
packaging
bottle of 100 mL
manufacturer/tradename
Tosoh 14479
parameter
≤50% organic solvent
3 bar max. pressure
technique(s)
LPLC: suitable
matrix
hydroxylated methacrylic polymer
matrix active group
phenyl phase
particle size
100 μm
operating pH
1-13
separation technique
hydrophobic interaction (HIC)
Looking for similar products? Visit Product Comparison Guide
General description
Specifications
Physical form
Legal Information
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Flam. Liq. 3
Storage Class Code
3 - Flammable liquids
WGK
WGK 3
Flash Point(F)
104.0 °F - closed cup
Flash Point(C)
40 °C - closed cup
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service