Skip to Content
Merck
All Photos(1)

Key Documents

76694

Supelco

2-Methylresorcinol

analytical standard

Synonym(s):

2,6-Dihydroxytoluene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3C6H3(OH)2
CAS Number:
Molecular Weight:
124.14
Beilstein:
2042177
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.21

grade

analytical standard

Quality Level

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

bp

264 °C (lit.)

mp

114-120 °C (lit.)

format

neat

storage temp.

2-30°C

SMILES string

Cc1c(O)cccc1O

InChI

1S/C7H8O2/c1-5-6(8)3-2-4-7(5)9/h2-4,8-9H,1H3

InChI key

ZTMADXFOCUXMJE-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Aquatic Acute 1 - Eye Dam. 1 - Skin Sens. 1B

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

275.0 °F - closed cup

Flash Point(C)

135 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service