Skip to Content
Merck
All Photos(1)

Key Documents

67661

Supelco

N-Nitrosoethylisopropylamine

analytical standard

Synonym(s):

N-Ethyl-N-nitroso-2-propanamine, EIPNA, NEIPA

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H12N2O
CAS Number:
Molecular Weight:
116.16
UNSPSC Code:
12352116
NACRES:
NA.05

grade

analytical standard

Quality Level

Assay

≥99%

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

format

neat

storage temp.

2-8°C

InChI

1S/C5H12N2O/c1-4-7(6-8)5(2)3/h5H,4H2,1-3H3

InChI key

VGGZTNNNXAUZLB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Carc. 1B

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

150.6 °F - (calculated)

Flash Point(C)

65.9 °C - (calculated)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service