20256-U
ORBO™ 70 Sorbent Tube
5% Na₂CO₃ on Chromosorb®-P (30/60), 335/165 mg, W,W,W separators, O.D. × L 8mm × 100mm, pkg of 50 ea
Synonym(s):
ORBO 70 Sorbent Tube, Chromosorb P, 30/60 mesh
About This Item
Recommended Products
product name
ORBO™ 70 5% Na2CO3 on Chromosorb®-P (30/60), 335/165 mg, W,W,W separators, O.D. × L 8 mm × 100 mm, pkg of 50 ea
material
W,W,W separators
Quality Level
product line
ORBO™
composition
Bed A, 335 mg
Bed B, 165 mg
packaging
pkg of 50 ea
manufacturer/tradename
ORBO™ 70
technique(s)
active air sampling: suitable
O.D. × L
8 mm × 100 mm
matrix
5% Na2 CO3 on Chromosorb-P
particle size
30-60 mesh
application(s)
air monitoring
environmental
industrial hygiene
Legal Information
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
STOT RE 1 Inhalation
Target Organs
Lungs
Storage Class Code
6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service