Z744062
SealPlate® RollMate™ Dispenser with1 Roll of SealPlate® Film
case of 1 ea (500 Pre-Scored Sheets)
All Photos(1)
About This Item
UNSPSC Code:
41122410
NACRES:
NC.01
Recommended Products
packaging
case of 1 ea (500 Pre-Scored Sheets)
manufacturer/tradename
Excel Scientific SP2-RL-500
parameter
-40 °C temp. range (120 °C)
technique(s)
ELISA: suitable
General description
- An economical, efficient way to rapidly seal microplates; faster than applying single sheeted film
- Self-standing, non-slip supports hold pre-scored film roll securely in place for quick and easy dispensing
- Continuous backing liner can be torn off or fed directly into a waste basket for disposal
- Roll tension can be easily adjusted to user preference (e.g. loose, tight)
- SealPlate film minimizes evaporation, reduces cross-contamination and prevents spillage
- 2.0 mil rigid polyester film features good clarity and easy handling
- Eliminates the “edge effect”, excess evaporation in the outside wells, which can occur with other films and plate lids
- Useful as a simple cover to protect samples from external contamination, dust and debris
- High integrity packaging prevents damage during transit; poly bag eliminates dust and particulate contamination
Legal Information
RollMate is a trademark of Excel Scientific, Inc.
SealPlate is a registered trademark of Excel Scientific, Inc.